Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tnip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199275.2 | NP_001186204.1 | 15 - 16 | 1861 | 85 | |
NM_001199276.2 | NP_001186205.1 | 13 - 14 | 1531 | 85 | ||
NM_001271455.1 | NP_001258384.1 | 14 - 15 | 1584 | 85 | ||
NM_001271456.1 | NP_001258385.1 | 15 - 16 | 1756 | 85 | ||
NM_021327.4 | NP_067302.2 | 14 - 15 | 1703 | 85 | ||
GenBank mRNA | AJ242777.1 | - | 13 - 14 | 1503 | 85 | |
AJ242778.1 | - | 14 - 15 | 1675 | 85 | ||
AK135338.1 | - | 13 - 14 | 1466 | 85 | ||
AK156131.1 | - | 14 - 15 | 1557 | 85 | ||
AK170841.1 | - | 13 - 14 | 1509 | 85 | ||
AK188071.1 | - | 3 - 4 | 307 | 85 | ||
AK193440.1 | - | 2 - 3 | 214 | 85 | ||
AK194753.1 | - | 3 - 4 | 276 | 85 | ||
AK217584.1 | - | 1 - 2 | 137 | 85 | ||
AY012159.2 | - | 11 - 12 | 1273 | 85 | ||
BC008186.1 | - | 14 - 15 | 1620 | 3589311 | 85 | |
BC008665.1 | - | 15 - 16 | 1727 | 3598934 | 85 | |
CB599739.1 | - | 5 - 6 | 590 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |