Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290805.1 | NP_001277734.1 | 7 - 8 | 1113 | 99 | |
NM_001290806.1 | NP_001277735.1 | 7 - 8 | 1113 | 99 | ||
NM_008443.4 | NP_032469.2 | 7 - 8 | 1113 | 99 | ||
GenBank mRNA | AK147078.1 | - | 7 - 8 | 1063 | 99 | |
AK161223.1 | - | 7 - 8 | 1104 | 99 | ||
AK161920.1 | - | 7 - 8 | 1087 | 99 | ||
AK169425.1 | - | 7 - 8 | 1087 | 99 | ||
BC023936.1 | - | 7 - 8 | 1063 | 99 | ||
BC044720.1 | - | 7 - 8 | 1074 | 99 | ||
BC052707.1 | - | 7 - 8 | 1079 | 5702415 | 99 | |
D12645.1 | - | 7 - 8 | 1089 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |