Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001166653.1 | NP_001160125.1 | 11 - 12 | 1837 | 152 | |
GenBank mRNA | AK166428.1 | - | 11 - 12 | 1772 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Kinase Mus musculus Probe spans exons |