Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mgat4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145926.3 | NP_666038.3 | 10 - 11 | 1456 | 60 | |
GenBank mRNA | AK155336.1 | - | 10 - 11 | 1457 | 60 | |
AK205818.1 | - | 1 - 2 | 62 | 60 | ||
AK211071.1 | - | 1 - 2 | 62 | 60 | ||
AK216064.1 | - | 1 - 2 | 62 | 60 | ||
BC026638.1 | - | 5 - 6 | 564 | 4984907 | 60 | |
BC031613.1 | - | 4 - 5 | 434 | 4216720 | 60 | |
CX205743.1 | - | 7 - 8 | 716 | 60 | ||
CX206051.1 | - | 2 - 3 | 215 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |