Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164056.1 | NP_001157528.1 | 25 - 26 | 3018 | 70 | |
NM_008875.4 | NP_032901.2 | 24 - 25 | 2963 | 70 | ||
GenBank mRNA | AK136111.1 | - | 20 - 21 | 2270 | 70 | |
BC026625.1 | - | 5 - 6 | 478 | 70 | ||
BC068144.1 | - | 24 - 25 | 2820 | 30532470 | 70 | |
U87868.1 | - | 25 - 26 | 3001 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |