Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrriq4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290510.1 | NP_001277439.1 | 4 - 5 | 1733 | 56 | |
NM_026668.2 | NP_080944.2 | 6 - 7 | 1896 | 56 | ||
GenBank mRNA | AK015024.1 | - | 4 - 5 | 1684 | 56 | |
AK019775.1 | - | 6 - 7 | 1897 | 56 | ||
BC145806.1 | - | 6 - 7 | 1597 | 40131109 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |