Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdh10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098170.1 | NP_001091640.1 | 2 - 3 | 3543 | 90 | |
NM_001098171.1 | NP_001091641.1 | 2 - 3 | 3543 | 90 | ||
NM_001098172.1 | NP_001091642.1 | 2 - 3 | 3543 | 90 | ||
GenBank mRNA | AF334801.1 | - | 2 - 3 | 2779 | 90 | |
AK034005.1 | - | 2 - 3 | 418 | 90 | ||
AK036195.1 | - | 2 - 3 | 1886 | 90 | ||
AK038615.1 | - | 2 - 3 | 1816 | 90 | ||
AK039226.1 | - | 2 - 3 | 889 | 90 | ||
AK046814.1 | - | 2 - 3 | 445 | 90 | ||
AK081621.1 | - | 2 - 3 | 3542 | 90 | ||
AK081977.1 | - | 2 - 3 | 3539 | 90 | ||
AK122503.1 | - | 2 - 3 | 3512 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |