Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcdh10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098170.1 | NP_001091640.1 | 3 - 4 | 3656 | 81 | |
NM_001098171.1 | NP_001091641.1 | 3 - 4 | 3656 | 81 | ||
NM_001098172.1 | NP_001091642.1 | 3 - 4 | 3656 | 81 | ||
GenBank mRNA | AF334801.1 | - | 3 - 4 | 2892 | 81 | |
AK034005.1 | - | 3 - 4 | 531 | 81 | ||
AK036195.1 | - | 3 - 4 | 1999 | 81 | ||
AK038615.1 | - | 3 - 4 | 1929 | 81 | ||
AK039226.1 | - | 3 - 4 | 1002 | 81 | ||
AK046814.1 | - | 3 - 4 | 558 | 81 | ||
AK081621.1 | - | 3 - 4 | 3655 | 81 | ||
AK081977.1 | - | 3 - 4 | 3652 | 81 | ||
AK122503.1 | - | 3 - 4 | 3625 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mus musculus Probe spans exons |