Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2ry14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001287121.1 | NP_001274050.1 | 1 - 2 | 323 | 93 | |
NM_001287123.1 | NP_001274052.1 | 1 - 2 | 323 | 93 | ||
GenBank mRNA | AK039791.1 | - | 1 - 2 | 325 | 93 | |
CF169635.1 | - | 1 - 2 | 208 | 93 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Mus musculus Inventoried Probe spans exons |