Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005509.2 | NP_001005509.1 | 12 - 13 | 1855 | 91 | |
GenBank mRNA | AK078519.1 | - | 12 - 13 | 1634 | 91 | |
AK145275.1 | - | 12 - 13 | 1812 | 91 | ||
BC030447.1 | - | 11 - 12 | 1524 | 5400637 | 91 | |
BC082557.1 | - | 12 - 13 | 1660 | 30606936 | 91 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |