Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Homer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001284189.1 | NP_001271118.1 | 7 - 8 | 1594 | 98 | |
NM_147176.3 | NP_671705.2 | 8 - 9 | 1630 | 98 | ||
NM_152134.3 | NP_687036.1 | 7 - 8 | 1113 | 98 | ||
GenBank mRNA | AB019479.1 | - | 8 - 9 | 1326 | 98 | |
AF093258.1 | - | 7 - 8 | 943 | 98 | ||
AK076543.1 | - | 2 - 3 | 163 | 98 | ||
AY137385.1 | - | 7 - 8 | 1113 | 98 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Probe spans exons |