Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sept8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252332.1 | NP_001239261.1 | 6 - 7 | 1063 | 75 | |
NM_001252333.1 | NP_001239262.1 | 6 - 7 | 1057 | 75 | ||
NM_033144.2 | NP_149156.1 | 6 - 7 | 1063 | 75 | ||
GenBank mRNA | AB093215.1 | - | 6 - 7 | 913 | 75 | |
AF179996.1 | - | 5 - 6 | 759 | 75 | ||
BC037707.1 | - | 6 - 7 | 851 | 75 | ||
BC049819.1 | - | 6 - 7 | 878 | 75 | ||
BC059248.1 | - | 6 - 7 | 999 | 6400429 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |