Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jkamp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205067.1 | NP_001191996.1 | 1 - 2 | 343 | 117 | |
GenBank mRNA | AK004555.1 | - | 1 - 2 | 298 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |