Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fcgr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077189.1 | NP_001070657.1 | 5 - 6 | 822 | 101 | |
GenBank mRNA | AK155329.1 | - | 5 - 6 | 101 | ||
AK171254.1 | - | 5 - 6 | 101 | |||
AK171316.1 | - | 5 - 6 | 805 | 101 | ||
M16367.1 | - | 7 - 8 | 1063 | 101 | ||
M17515.1 | - | 5 - 6 | 776 | 101 | ||
U31801.1 | - | 3 - 4 | 637 | 101 | ||
U31802.1 | - | 3 - 4 | 101 | |||
U51629.1 | - | 5 - 6 | 730 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |