Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scyl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001286002.1 | NP_001272931.1 | 2 - 3 | 448 | 117 | |
NM_028776.5 | NP_083052.1 | 2 - 3 | 635 | 117 | ||
GenBank mRNA | AK004809.2 | - | 2 - 3 | 375 | 117 | |
AK030824.1 | - | 2 - 3 | 611 | 117 | ||
AK162937.1 | - | 2 - 3 | 449 | 117 | ||
BC043085.1 | - | 2 - 3 | 336 | 6417440 | 117 | |
BC066800.1 | - | 2 - 3 | 320 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |