Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010880.3 | NP_035010.3 | 9 - 10 | 1684 | 86 | |
GenBank mRNA | AF318184.1 | - | 9 - 10 | 1596 | 86 | |
AK028681.1 | - | 9 - 10 | 1594 | 86 | ||
AK029128.1 | - | 9 - 10 | 1594 | 86 | ||
AK031606.1 | - | 9 - 10 | 1630 | 86 | ||
AK050958.1 | - | 7 - 8 | 981 | 86 | ||
AK083307.1 | - | 9 - 10 | 1604 | 86 | ||
AK088402.1 | - | 9 - 10 | 1595 | 86 | ||
AK144894.1 | - | 9 - 10 | 1594 | 86 | ||
AK161597.1 | - | 9 - 10 | 1594 | 86 | ||
AK161706.1 | - | 9 - 10 | 1604 | 86 | ||
AK163275.1 | - | 9 - 10 | 1595 | 86 | ||
AK166681.1 | - | 9 - 10 | 1594 | 86 | ||
AK168579.1 | - | 9 - 10 | 1594 | 86 | ||
AK168623.1 | - | 9 - 10 | 1594 | 86 | ||
BC005460.1 | - | 9 - 10 | 1504 | 3495665 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons |