Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mnat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008612.2 | NP_032638.2 | 3 - 4 | 432 | 153 | |
GenBank mRNA | AK008249.1 | - | 3 - 4 | 435 | 153 | |
AK029174.1 | - | 3 - 4 | 344 | 153 | ||
AK155859.1 | - | 3 - 4 | 433 | 153 | ||
AK157721.1 | - | 3 - 4 | 419 | 153 | ||
BC089023.1 | - | 3 - 4 | 421 | 30702121 | 153 | |
BC115630.2 | - | 3 - 4 | 424 | 40102880 | 153 | |
BC115631.1 | - | 3 - 4 | 425 | 153 | ||
U35249.1 | - | 3 - 4 | 463 | 153 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |