Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178405.3 | NP_848492.1 | 6 - 7 | 796 | 69 | |
GenBank mRNA | AK129212.1 | - | 6 - 7 | 709 | 69 | |
AK137602.1 | - | 6 - 7 | 732 | 69 | ||
AK147331.1 | - | 6 - 7 | 728 | 69 | ||
AK148775.1 | - | 6 - 7 | 728 | 69 | ||
AK161840.1 | - | 6 - 7 | 728 | 69 | ||
AK164509.1 | - | 6 - 7 | 732 | 69 | ||
AK168703.1 | - | 6 - 7 | 728 | 69 | ||
BC036127.1 | - | 6 - 7 | 720 | 4955003 | 69 | |
BC041774.1 | - | 3 - 4 | 446 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Ion Channel Transporter Mammalian Gene Collection Mus musculus Probe spans exons |