Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sox13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011439.2 | NP_035569.2 | 7 - 8 | 1094 | 154 | |
GenBank mRNA | AB006329.1 | - | 6 - 7 | 750 | 154 | |
AJ000740.1 | - | 6 - 7 | 154 | |||
BC007130.1 | - | 7 - 8 | 3493465 | 154 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |