Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009791.4 | NP_033921.3 | 12 - 13 | 3234 | 137 | |
GenBank mRNA | AF533752.1 | - | 12 - 13 | 3212 | 137 | |
AK052326.1 | - | 11 - 12 | 3323 | 137 | ||
AY971958.1 | - | 12 - 13 | 3075 | 137 | ||
CN664812.1 | - | 3 - 4 | 388 | 137 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |