Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aspm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009791.4 | NP_033921.3 | 1 - 2 | 427 | 82 | |
GenBank mRNA | AF533752.1 | - | 1 - 2 | 405 | 82 | |
AK050785.1 | - | 1 - 2 | 423 | 82 | ||
AK052326.1 | - | 1 - 2 | 428 | 82 | ||
AK079744.1 | - | 1 - 2 | 393 | 82 | ||
AK083710.1 | - | 1 - 2 | 394 | 82 | ||
AY971958.1 | - | 1 - 2 | 268 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |