Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100010814.mm.1 |
NM_009791.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010814.mm.1 |
NM_009791.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010814.mm.1 |
TC0100001303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100010813.mm.1 |
NM_009791.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010813.mm.1 |
NM_009791.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010813.mm.1 |
TC0100001303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100010817.mm.1 |
NM_009791.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010817.mm.1 |
NM_009791.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010817.mm.1 |
TC0100001303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100010816.mm.1 |
NM_009791.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010816.mm.1 |
NM_009791.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010816.mm.1 |
TC0100001303.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100010815.mm.1 |
NM_009791.4.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010815.mm.1 |
NM_009791.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100010815.mm.1 |
TC0100001303.mm.1 |
MTA TC |