Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AY513272.1 | - | 9 - 10 | 1150 | 151 | |
BC005581.1 | - | 9 - 10 | 1123 | 3968615 | 151 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |