Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tor1aip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160018.1 | NP_001153490.1 | 5 - 6 | 943 | 142 | |
NM_001160019.1 | NP_001153491.1 | 5 - 6 | 357 | 142 | ||
NM_144791.2 | NP_659040.2 | 5 - 6 | 943 | 142 | ||
GenBank mRNA | AB251963.1 | - | 5 - 6 | 883 | 142 | |
AK149727.1 | - | 5 - 6 | 863 | 142 | ||
AK152751.1 | - | 5 - 6 | 377 | 142 | ||
BC010841.1 | - | 5 - 6 | 758 | 3493883 | 142 | |
BQ891636.1 | - | 5 - 6 | 538 | 142 | ||
CA465752.1 | - | 5 - 6 | 360 | 142 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |