Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fasl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010177.4 | NP_034307.1 | 1 - 2 | 556 | 120 | |
GenBank mRNA | AK040139.1 | - | 1 - 2 | 556 | 120 | |
AK141268.1 | - | 1 - 2 | 556 | 120 | ||
BC052866.1 | - | 653 | 30073084 | 120 | ||
S76752.1 | - | 1 - 2 | 376 | 120 | ||
U06948.1 | - | 1 - 2 | 470 | 120 | ||
U10984.1 | - | 1 - 2 | 526 | 120 | ||
U58995.1 | - | 1 - 2 | 346 | 120 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Mus musculus 1700 Probe spans exons |