Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ncstn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021607.3 | NP_067620.3 | 12 - 13 | 1519 | 58 | |
GenBank mRNA | AF240469.1 | - | 12 - 13 | 1469 | 58 | |
AK033314.1 | - | 12 - 13 | 1462 | 58 | ||
AK129102.1 | - | 12 - 13 | 1519 | 58 | ||
AK160338.1 | - | 65 | 58 | |||
AK170244.1 | - | 12 - 13 | 1484 | 58 | ||
AK170369.1 | - | 12 - 13 | 1508 | 58 | ||
AK172579.1 | - | 12 - 13 | 1464 | 58 | ||
BC019998.1 | - | 12 - 13 | 1464 | 4458780 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |