Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arl8a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026823.2 | NP_081099.1 | 2 - 3 | 352 | 110 | |
GenBank mRNA | AB185207.1 | - | 2 - 3 | 208 | 110 | |
BC004035.1 | - | 1 - 2 | 83 | 110 | ||
BC018479.1 | - | 2 - 3 | 352 | 3661329 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |