Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sypl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013635.3 | NP_038663.2 | 1 - 2 | 80 | 67 | |
GenBank mRNA | BC060971.1 | - | 1 - 2 | 80 | 6770686 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons |