Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mamld1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081354.2 | NP_001074823.2 | 3 - 4 | 2166 | 85 | |
GenBank mRNA | BC157923.1 | - | 2 - 3 | 2079 | 9088096 | 85 |
BC172017.1 | - | 2 - 3 | 2094 | 9054303 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |