Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc26a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177243.4 | NP_796217.2 | 1 - 2 | 87 | 86 | |
GenBank mRNA | AK078966.1 | - | 1 - 2 | 88 | 86 | |
AK086815.1 | - | 1 - 2 | 88 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Transporter Mus musculus Probe spans exons |