Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_172049.2 | NP_742046.2 | 1 - 2 | 122 | 88 | |
GenBank mRNA | AK133493.1 | - | 1 - 2 | 122 | 88 | |
AK160817.1 | - | 1 - 2 | 114 | 88 | ||
AK166323.1 | - | 1 - 2 | 139 | 88 | ||
AK170496.1 | - | 1 - 2 | 105 | 88 | ||
BC024093.1 | - | 1 - 2 | 94 | 5116657 | 88 | |
BY013255.1 | - | 1 - 2 | 123 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |