Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Itm2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022417.3 | NP_071862.2 | 1 - 2 | 275 | 77 | |
GenBank mRNA | AB030199.1 | - | 1 - 2 | 218 | 77 | |
AF272044.1 | - | 1 - 2 | 182 | 77 | ||
AF282981.1 | - | 1 - 2 | 203 | 77 | ||
AK036377.1 | - | 1 - 2 | 221 | 77 | ||
AK156930.1 | - | 1 - 2 | 178 | 77 | ||
BC012952.1 | - | 1 - 2 | 153 | 2648812 | 77 | |
BY255015.1 | - | 1 - 2 | 277 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |