Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P551844 |
BC096763.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P551844 |
AK079810.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P551844 |
BC096763.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P551844 |
AK079810.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P429364 |
AK170686.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P429364 |
BC096763.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P429364 |
NM_177471.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P429364 |
AK008760.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P429364 |
AK135942.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P429364 |
AK170686.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P429364 |
BC096763.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P429364 |
NM_177471.4 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P429364 |
AK008760.1 |
mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_52_P429364 |
AK135942.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
235173 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
952682 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
793711 |
AK013105.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
793711 |
NM_026515.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
962017 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
33204 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
897190 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
316646 |
AK031954.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
316646 |
AK044068.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
316646 |
AK078098.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
316646 |
AK138778.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
316646 |
BC079890.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
316646 |
NM_172946.2 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
758084 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
799176 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
513917 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
761292 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
709759 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
283440 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
10442 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
596678 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
577924 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
929394 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
97033 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
52971 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
48487 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P166405 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P151837 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P429364 |
AK170686.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P429364 |
BC096763.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P429364 |
NM_177471.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P429364 |
AK008760.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P429364 |
AK135942.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P429364 |
AK170686.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P429364 |
BC096763.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P429364 |
NM_177471.4 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P429364 |
AK008760.1 |
mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_52_P429364 |
AK135942.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27813599 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27813587 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27813585 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27813591 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27813583 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27813592 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27813608 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27813594 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27813598 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27813597 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27813604 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27813581 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27813595 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27813584 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27813590 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27813593 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27813603 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27813601 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27813606 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27813610 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27813602 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P02330020 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P166405 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P151837 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07349444 |
- |
- |
Whole Mouse Genome G4122A |
A_52_P551844 |
BC096763.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P551844 |
AK079810.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P551844 |
BC096763.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P551844 |
AK079810.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P429364 |
AK170686.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P429364 |
BC096763.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P429364 |
NM_177471.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P429364 |
AK008760.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P429364 |
AK135942.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P429364 |
AK170686.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P429364 |
BC096763.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P429364 |
NM_177471.4 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P429364 |
AK008760.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_52_P429364 |
AK135942.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P118398 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P166405 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P151837 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09075678 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09188106 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09509413 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813629 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813540 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813626 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813436 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813618 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813578 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813592 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813525 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813524 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813569 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813632 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813583 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813594 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813432 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813492 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813494 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813510 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813617 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813565 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813438 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813581 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813621 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813499 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813481 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813488 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813610 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813620 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813630 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813501 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813478 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813437 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813577 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813586 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813604 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813538 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813609 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813624 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813541 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813590 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813536 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813596 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813509 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813606 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813464 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813519 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813449 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813603 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813441 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813622 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813572 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813521 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813451 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813591 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813518 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813467 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813529 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813484 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813556 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813534 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813470 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813571 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813514 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813483 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813584 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813486 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813522 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813465 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813454 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813461 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813573 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813537 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813532 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813503 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813598 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813459 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813527 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813557 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813566 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813550 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813507 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813520 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813602 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813548 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813553 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813477 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813561 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813496 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813568 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813545 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27813547 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813620 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813581 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813621 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09220799 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813617 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813577 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813583 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09110584 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813550 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813610 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09411449 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813613 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09549947 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813603 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09154779 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813553 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09537298 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09502933 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09110582 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09509413 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813614 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813584 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813570 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813606 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09110585 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813596 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813601 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813573 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09209100 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813579 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813569 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813565 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813586 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813623 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813600 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813602 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813571 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09144558 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813592 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09188108 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813618 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813563 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813590 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813575 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813568 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813578 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813608 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813574 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813597 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27813598 |
- |
- |