Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cttnbp2nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163332.1 | NP_001156804.1 | 2 - 3 | 179 | 90 | |
NM_030249.4 | NP_084525.1 | 2 - 3 | 287 | 90 | ||
GenBank mRNA | AK029901.1 | - | 2 - 3 | 288 | 90 | |
AK030438.1 | - | 2 - 3 | 152 | 90 | ||
AK051259.1 | - | 2 - 3 | 151 | 90 | ||
AK129359.1 | - | 2 - 3 | 177 | 90 | ||
AK145085.1 | - | 2 - 3 | 150 | 90 | ||
BC003236.1 | - | 2 - 3 | 141 | 3498078 | 90 | |
BY195820.1 | - | 2 - 3 | 184 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |