Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmnl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_011711.1 | NP_035841.1 | 8 - 9 | 959 | 61 | |
GenBank mRNA | AK129473.1 | - | 8 - 9 | 878 | 61 | |
AK139711.1 | - | 8 - 9 | 942 | 61 | ||
AK143308.1 | - | 8 - 9 | 858 | 61 | ||
AK155331.1 | - | 8 - 9 | 976 | 61 | ||
AK163361.1 | - | 8 - 9 | 945 | 61 | ||
AK170744.1 | - | 7 - 8 | 789 | 61 | ||
BC023797.1 | - | 7 - 8 | 679 | 61 | ||
BC085094.1 | - | 8 - 9 | 856 | 61 | ||
BC131961.1 | - | 8 - 9 | 835 | 40130238 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |