Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025730.3 | NP_080006.3 | 42 - 43 | 6335 | 149 | |
GenBank mRNA | AK014886.1 | - | 3 - 4 | 394 | 149 | |
AK014938.1 | - | 3 - 4 | 407 | 149 | ||
AY792512.1 | - | 42 - 43 | 6278 | 149 | ||
BC034074.1 | - | 8 - 9 | 1218 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |