Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cntnap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081129.1 | NP_001074598.1 | 15 - 16 | 2414 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mus musculus Probe spans exons |