Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aatf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019816.1 | NP_062790.1 | 4 - 5 | 719 | 123 | |
GenBank mRNA | AF222801.1 | - | 4 - 5 | 719 | 123 | |
AK077789.1 | - | 4 - 5 | 743 | 123 | ||
AK141334.1 | - | 4 - 5 | 717 | 123 | ||
BC025080.1 | - | 4 - 5 | 706 | 2648117 | 123 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |