Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080470.1 | NP_536718.1 | 15 - 16 | 2429 | 119 | |
GenBank mRNA | AF303827.1 | - | 15 - 16 | 2429 | 119 | |
BC129800.1 | - | 15 - 16 | 2420 | 40048794 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |