Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rictor | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030168.3 | NP_084444.3 | 22 - 23 | 2201 | 89 | |
GenBank mRNA | AK036149.1 | - | 22 - 23 | 2202 | 89 | |
AK082586.1 | - | 22 - 23 | 2192 | 89 | ||
AY497009.1 | - | 22 - 23 | 2179 | 89 | ||
AY540053.1 | - | 22 - 23 | 2199 | 89 | ||
BC022906.1 | - | 17 - 18 | 1747 | 89 | ||
BC120903.1 | - | 18 - 19 | 1840 | 40086992 | 89 | |
BC120904.1 | - | 22 - 23 | 2199 | 40086995 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |