Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atad5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029856.2 | NP_001025027.1 | 11 - 12 | 3565 | 117 | |
GenBank mRNA | AY557610.1 | - | 11 - 12 | 3556 | 117 | |
BC046463.1 | - | 7 - 8 | 862 | 117 | ||
BC141180.1 | - | 10 - 11 | 3061 | 9055792 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |