Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mllt6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_139311.2 | NP_647472.2 | 5 - 6 | 616 | 53 | |
GenBank mRNA | AK143477.1 | - | 1 - 2 | 59 | 53 | |
AY050217.1 | - | 5 - 6 | 763 | 53 | ||
BY731745.1 | - | 1 - 2 | 55 | 53 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |