Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pcgf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163307.1 | NP_001156779.1 | 1 - 2 | 175 | 87 | |
NM_009545.2 | NP_033571.1 | 3 - 4 | 442 | 87 | ||
GenBank mRNA | BC016419.1 | - | 1 - 2 | 129 | 4458285 | 87 |
D90085.1 | - | 3 - 4 | 290 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |