Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttc23l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029430.1 | NP_083706.1 | 8 - 9 | 1078 | 74 | |
GenBank mRNA | AK015027.1 | - | 8 - 9 | 1050 | 74 | |
BC132384.1 | - | 8 - 9 | 1078 | 74 | ||
BC145852.1 | - | 8 - 9 | 1078 | 40131155 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |