Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Axin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009733.2 | NP_033863.2 | 8 - 9 | 2625 | 81 | |
GenBank mRNA | AF009011.1 | - | 8 - 9 | 2577 | 81 | |
BC011544.1 | - | 8 - 9 | 2344 | 81 | ||
BC069954.1 | - | 1 - 2 | 107 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |