Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdac4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_207225.1 | NP_997108.1 | 7 - 8 | 980 | 104 | |
GenBank mRNA | AK029933.1 | - | 4 - 5 | 517 | 104 | |
AK155250.1 | - | 8 - 9 | 1166 | 104 | ||
BC066052.1 | - | 7 - 8 | 980 | 6827645 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |