Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177642.3 | NP_808310.2 | 23 - 24 | 7715 | 75 | |
GenBank mRNA | BC107027.1 | - | 1 - 2 | 39 | 40050318 | 75 |
BC107028.2 | - | 1 - 2 | 39 | 40050319 | 75 | |
EU429481.1 | - | 9 - 10 | 4587 | 75 | ||
EU429482.1 | - | 8 - 9 | 1454 | 75 | ||
EU429483.2 | - | 18 - 19 | 2868 | 75 | ||
GU290042.1 | - | 23 - 24 | 7715 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |