Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rpl3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163945.1 | NP_001157417.1 | 5 - 6 | 742 | 93 | |
NM_025425.4 | NP_079701.1 | 2 - 3 | 409 | 93 | ||
GenBank mRNA | AK002393.1 | - | 2 - 3 | 286 | 93 | |
AK004280.1 | - | 2 - 3 | 410 | 93 | ||
BC085243.1 | - | 2 - 3 | 310 | 93 | ||
BC100562.1 | - | 2 - 3 | 255 | 93 | ||
BC145876.1 | - | 2 - 3 | 235 | 40131179 | 93 | |
BC145878.1 | - | 2 - 3 | 235 | 40131181 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |