Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nr1d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145434.4 | NP_663409.2 | 2 - 3 | 999 | 75 | |
GenBank mRNA | AF291821.1 | - | 2 - 3 | 823 | 75 | |
AK041047.1 | - | 2 - 3 | 854 | 75 | ||
AK135640.1 | - | 2 - 3 | 861 | 75 | ||
AK137398.1 | - | 2 - 3 | 886 | 75 | ||
AK137582.1 | - | 2 - 3 | 864 | 75 | ||
AK146430.1 | - | 2 - 3 | 956 | 75 | ||
AK154931.1 | - | 2 - 3 | 1000 | 75 | ||
AK155597.1 | - | 2 - 3 | 1000 | 75 | ||
BC008989.1 | - | 2 - 3 | 444 | 3585478 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Nuclear Receptor Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |