Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sgca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009161.4 | NP_033187.1 | 6 - 7 | 829 | 64 | |
GenBank mRNA | AB024920.1 | - | 6 - 7 | 829 | 64 | |
AK009015.1 | - | 5 - 6 | 592 | 64 | ||
AK075915.1 | - | 5 - 6 | 601 | 64 | ||
BC060287.1 | - | 5 - 6 | 588 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |